Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021702.1 | NP_067734.1 | 7 | 611 | ||
XM_006240485.2 | XP_006240547.1 | 8 | 647 | |||
XM_006240486.2 | XP_006240548.1 | 8 | 644 | |||
XM_006240487.2 | XP_006240549.1 | 8 | 647 | |||
XM_006240488.2 | XP_006240550.1 | 7 | 563 | |||
XM_006240490.1 | XP_006240552.1 | 8 | 673 | |||
XM_006240493.2 | XP_006240555.1 | 7 | 566 | |||
GenBank mRNA | Y12319.1 | - | 7 | 611 |