Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004131.4 | NP_004122.2 | 4 | 578 | ||
XM_011536685.2 | XP_011534987.1 | 4 | 562 | |||
GenBank mRNA | AY232654.1 | - | 4 | 543 | ||
AY232655.1 | - | 3 | 380 | |||
AY232656.1 | - | 2 | 233 | |||
AY372494.1 | - | 4 | 619 | |||
BC030195.1 | - | 4 | 534 | 5223876 | ||
J03189.1 | - | 4 | 551 | |||
J04071.1 | - | 4 | 528 | |||
M17016.1 | - | 4 | 519 |