Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parva | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020656.2 | NP_065707.2 | 7 | 704 | ||
GenBank mRNA | AF264765.2 | - | 7 | 704 | ||
FQ228521.1 | - | 764 |