Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021589.1 | NP_067600.1 | 14 | 1719 | ||
XM_006232746.2 | XP_006232808.1 | 13 | 1692 | |||
GenBank mRNA | M85214.1 | - | 14 | 1719 |