Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021597.1 | NP_067608.1 | 5 | 784 | ||
GenBank mRNA | AF195534.1 | - | 5 | 784 |