Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBAG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278938.1 | NP_001265867.1 | 4 | 346 | ||
NM_004215.4 | NP_004206.1 | 4 | 633 | |||
NM_198120.2 | NP_936056.1 | 4 | 466 | |||
XM_017013960.1 | XP_016869449.1 | 4 | 829 | |||
GenBank mRNA | AB007619.1 | - | 4 | 514 | ||
AF006265.1 | - | 4 | 464 | |||
AK290651.1 | - | 4 | 455 | |||
AK308950.1 | - | 4 | 524 | |||
AL515533.3 | - | 4 | 524 | |||
AY515724.1 | - | 4 | 424 | |||
AY653072.1 | - | 3 | 231 | |||
BC005249.1 | - | 4 | 544 | 3930583 | ||
BC017729.1 | - | 4 | 417 | 3453385 | ||
BC022506.1 | - | 4 | 531 | 4815654 | ||
BG616704.1 | - | 4 | 347 | |||
BG707115.1 | - | 4 | 467 | |||
BI194881.1 | - | 4 | 633 | |||
CD108096.1 | - | 4 | 577 | |||
CR456984.1 | - | 3 | 231 |