Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021740.1 | NP_068508.1 | 3 | 310 | ||
GenBank mRNA | BC061972.1 | - | 3 | 320 | 5623463 | |
BC092569.1 | - | 3 | 330 | 7307282 | ||
FQ210626.1 | - | 3 | 341 | |||
FQ212611.1 | - | 3 | 341 | |||
FQ214221.1 | - | 3 | 335 | |||
FQ215633.1 | - | 3 | 341 | |||
FQ219686.1 | - | 3 | 341 | |||
FQ220677.1 | - | 3 | 342 | |||
FQ225103.1 | - | 3 | 345 | |||
FQ226916.1 | - | 3 | 341 | |||
FQ226946.1 | - | 3 | 341 | |||
FQ227002.1 | - | 3 | 341 | |||
FQ227835.1 | - | 3 | 341 | |||
FQ229930.1 | - | 3 | 341 | |||
FQ231540.1 | - | 3 | 341 | |||
FQ231794.1 | - | 3 | 347 | |||
FQ233285.1 | - | 3 | 347 | |||
M20035.1 | - | 3 | 337 | |||
M33024.1 | - | 3 | 337 | |||
M60664.1 | - | 3 | 349 | |||
M60665.1 | - | 3 | 349 | |||
M86564.1 | - | 3 | 310 | |||
X55326.1 | - | 3 | 310 |
Gene Symbol: LOC100359583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751278.3 | XP_003751326.1 | 1 | 347 |
Gene Symbol: LOC680837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769073.1 | XP_008767295.1 | 1 | 164 |