Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021754.1 | NP_068522.1 | 11, 12 | 1381 | ||
XM_006245013.1 | XP_006245075.1 | 10, 11 | 1206 | |||
GenBank mRNA | AF194371.1 | - | 11, 12 | 1381 | ||
BC087637.1 | - | 11, 12 | 1414 | 7309062 | ||
FQ225580.1 | - | 4, 5 | 577 | |||
FQ230388.1 | - | 4, 5 | 577 |