Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021846.2 | NP_068618.2 | 736 | |||
GenBank mRNA | AF115380.1 | - | 668 | |||
BC078835.1 | - | 736 | 7121699 |