Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021857.3 | NP_068629.2 | 1 | 329 |
Set Membership: |
G-protein coupled receptor |