Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022190.1 | NP_071526.1 | 3 | 636 | ||
GenBank mRNA | J03485.1 | - | 3 | 636 |