Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022193.1 | NP_071529.1 | 1 | 209 | ||
GenBank mRNA | EF121986.1 | - | 1 | 209 | ||
EF121987.1 | - | 1 | 209 | |||
J03808.1 | - | 1 | 209 |