Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022205.3 | NP_071541.2 | 2 | 283 | ||
GenBank mRNA | AF452185.1 | - | 1 | 208 | ||
BC089804.1 | - | 2 | 263 | 7380432 | ||
CO557829.1 | - | 2 | 283 | |||
U54791.1 | - | 2 | 251 | |||
U90610.1 | - | 1 | 208 |