Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022207.1 | NP_071543.1 | 2 | 173 | ||
XM_006256423.2 | XP_006256485.1 | 2 | 569 | |||
GenBank mRNA | U87306.1 | - | 2 | 173 |