Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021759.1 | NP_068527.1 | 3, 4 | 476 | ||
GenBank mRNA | AJ001043.1 | - | 3, 4 | 476 |