Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271696.1 | NP_001258625.1 | 2 | 299 | ||
NM_001271697.1 | NP_001258626.1 | 2 | 299 | |||
NM_001271699.1 | NP_001258628.1 | 2 | 302 | |||
NM_004299.4 | NP_004290.2 | 2 | 302 | |||
GenBank mRNA | AF038950.1 | - | 2 | 239 | ||
AF078777.1 | - | 2 | 219 | |||
AF133659.1 | - | 2 | 231 | |||
AK001418.1 | - | 2 | 259 | |||
AK307414.1 | - | 2 | 222 | |||
BC006323.2 | - | 2 | 235 | 4138236 | ||
BG722908.1 | - | 2 | 277 | |||
BP228998.1 | - | 2 | 302 | |||
BT009918.1 | - | 2 | 234 | |||
BX537833.1 | - | 2 | 238 | |||
KU177872.1 | - | 2 | 234 | |||
KU177873.1 | - | 2 | 234 | |||
KU177875.1 | - | 2 | 231 |