Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021851.1 | NP_068623.1 | 3, 4 | 236 | ||
GenBank mRNA | AF090136.1 | - | 3, 4 | 236 | ||
FQ212254.1 | - | 268 |