Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021867.3 | NP_068639.1 | 3 | 527 | ||
GenBank mRNA | AB002561.1 | - | 3 | 527 | ||
BC085738.1 | - | 801 | 7189721 |