Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022175.1 | NP_071511.1 | 2 | 201 | ||
XM_006257472.2 | XP_006257534.1 | 3 | 650 | |||
XM_006257473.2 | XP_006257535.1 | 3 | 266 | |||
XM_006257475.1 | XP_006257537.1 | 4 | 394 | |||
XM_008773400.1 | XP_008771622.1 | 4 | 545 | |||
GenBank mRNA | DQ058653.1 | - | 2 | 201 | ||
FQ228987.1 | - | 3 | 284 | |||
S82627.1 | - | 2 | 186 |