Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022212.2 | NP_071548.2 | 4 | 1393 | ||
XM_006232749.2 | XP_006232811.2 | 4 | 1463 | |||
XM_008761207.1 | XP_008759429.1 | 4 | 1463 | |||
GenBank mRNA | D13965.1 | - | 4 | 995 | ||
D13966.1 | - | 4 | 995 | |||
M90661.1 | - | 4 | 1393 |