Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021851.1 | NP_068623.1 | 4 | 336 | ||
GenBank mRNA | AF090136.1 | - | 4 | 336 | ||
FQ212254.1 | - | 368 |