Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022184.1 | NP_071520.1 | 13, 14 | 1598 | ||
GenBank mRNA | U47110.1 | - | 13, 14 | 1598 |