Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022245.1 | NP_071581.1 | 1 | 174 | ||
GenBank mRNA | AF007107.1 | - | 1 | 137 | ||
AF007108.1 | - | 1 | 137 | |||
BC086945.1 | - | 1 | 168 | 7322141 | ||
D13205.1 | - | 1 | 174 | |||
FQ209861.1 | - | 1 | 165 | |||
FQ210790.1 | - | 1 | 162 | |||
FQ224354.1 | - | 1 | 165 |