Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PVALB SIRNA Assays ›
Gene Symbol:
Pvalb
Gene Name:
parvalbumin
Gene Aliases:
PALB1, Pva
Chromosome Location:
Chr.7: 119428652 - 119443656 on Build Rnor_6.0
UniGene ID:
Rn.2005
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
51787
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pvalb
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022499.2
NP_071944.1
2
158
XM_006241929.2
XP_006241991.1
3
231
GenBank mRNA
BC126090.1
-
2
158
7321065
FQ212120.1
-
3
221
FQ214597.1
-
3
219
FQ214667.1
-
3
220
FQ214909.1
-
3
219
FQ215072.1
-
3
219
FQ215101.1
-
1
55
FQ215172.1
-
3
219
FQ215177.1
-
3
218
FQ215250.1
-
3
220
FQ215353.1
-
3
219
FQ215367.1
-
3
219
FQ215388.1
-
3
219
FQ215455.1
-
3
219
FQ215472.1
-
3
219
FQ215487.1
-
3
219
FQ215608.1
-
3
220
FQ215681.1
-
3
219
FQ216147.1
-
3
219
FQ216186.1
-
3
219
FQ216201.1
-
3
220
FQ216260.1
-
3
219
FQ216338.1
-
3
220
FQ216339.1
-
3
219
FQ216448.1
-
3
219
FQ216459.1
-
3
219
FQ216533.1
-
3
219
FQ216659.1
-
3
219
FQ216789.1
-
3
219
FQ216830.1
-
3
219
FQ216843.1
-
3
219
FQ216867.1
-
3
219
FQ216872.1
-
3
219
FQ216903.1
-
3
221
FQ216906.1
-
3
216
FQ216960.1
-
3
219
FQ216981.1
-
3
219
FQ217053.1
-
3
219
FQ217064.1
-
3
219
FQ217099.1
-
3
219
FQ217132.1
-
2
220
FQ217209.1
-
3
218
FQ217210.1
-
3
219
FQ217278.1
-
3
219
FQ217298.1
-
3
220
FQ217373.1
-
3
220
FQ217384.1
-
3
219
FQ217418.1
-
3
220
FQ217424.1
-
3
219
FQ217446.1
-
2
187
FQ217489.1
-
3
219
FQ217527.1
-
3
219
FQ217562.1
-
3
219
FQ217576.1
-
3
219
FQ217645.1
-
3
218
FQ217652.1
-
3
220
FQ217677.1
-
3
220
FQ217743.1
-
3
218
FQ217749.1
-
3
220
FQ217760.1
-
3
220
FQ217770.1
-
3
219
FQ217787.1
-
3
220
FQ217845.1
-
3
219
FQ217858.1
-
3
219
FQ217860.1
-
3
220
FQ217861.1
-
3
219
FQ217880.1
-
3
221
FQ217886.1
-
3
219
FQ217934.1
-
2
161
FQ217966.1
-
3
219
FQ217976.1
-
3
219
FQ217991.1
-
3
219
FQ218022.1
-
3
219
FQ218035.1
-
3
219
FQ218057.1
-
3
219
FQ223582.1
-
3
219
FQ223596.1
-
3
219
FQ223603.1
-
3
220
FQ223630.1
-
3
216
FQ223664.1
-
3
220
FQ223669.1
-
1
53
FQ223695.1
-
3
219
FQ223712.1
-
3
220
FQ223861.1
-
3
219
FQ223863.1
-
3
219
FQ223909.1
-
3
219
FQ223941.1
-
3
219
FQ223961.1
-
3
219
FQ223988.1
-
3
218
FQ223989.1
-
3
219
FQ223990.1
-
3
219
FQ224014.1
-
3
219
FQ224025.1
-
3
220
FQ224028.1
-
3
219
FQ224037.1
-
3
219
FQ224053.1
-
3
219
FQ224059.1
-
3
219
FQ224083.1
-
3
219
FQ224113.1
-
3
219
FQ224136.1
-
3
219
FQ224173.1
-
3
219
FQ224181.1
-
3
219
FQ224242.1
-
3
220
FQ224248.1
-
3
217
FQ224294.1
-
3
219
FQ224303.1
-
3
220
FQ224322.1
-
3
219
FQ224356.1
-
3
219
FQ224363.1
-
3
219
FQ224417.1
-
3
219
FQ224430.1
-
3
219
FQ224435.1
-
3
217
FQ224463.1
-
3
220
FQ224483.1
-
3
219
FQ224485.1
-
3
219
FQ224488.1
-
3
220
FQ224510.1
-
3
219
FQ224541.1
-
3
219
FQ224580.1
-
3
219
FQ224584.1
-
3
220
FQ224600.1
-
3
219
FQ224613.1
-
3
219
FQ224634.1
-
3
219
FQ224643.1
-
3
219
FQ224673.1
-
3
220
FQ224675.1
-
3
219
FQ224723.1
-
3
221
FQ224767.1
-
3
219
FQ224778.1
-
3
219
FQ224913.1
-
3
219
M10764.1
-
1
44
M12725.1
-
3
218
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytosolic calcium ion concentration
calcium ion binding
protein homodimerization activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline