Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022281.2 | NP_071617.2 | 4 | 453 | ||
GenBank mRNA | AF487549.1 | - | 4 | 453 | ||
AY170916.1 | - | 4 | 414 | |||
AY174892.1 | - | 4 | 453 | |||
AY174893.1 | - | 4 | 453 |
Gene Symbol: LOC103693258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767460.1 | XP_008765682.1 | 3 | 894 |
Gene Symbol: LOC688708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767477.1 | XP_008765699.1 | 3 | 894 |