Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022289.3 | NP_071625.2 | 2 | 470 | ||
XM_006232135.2 | XP_006232197.1 | 2 | 483 | |||
GenBank mRNA | AF068746.2 | - | 1 | 182 | ||
AF305780.1 | - | 2 | 456 | |||
BC061554.1 | - | 2 | 474 | 6890629 |