Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022302.1 | NP_071638.1 | 2 | 190 | ||
GenBank mRNA | AF193755.1 | - | 2 | 190 |