Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022380.1 | NP_071775.1 | 274 | |||
GenBank mRNA | X91988.1 | - | 2 | 308 | ||
X97541.1 | - | 274 |