Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022403.2 | NP_071798.1 | 5 | 355 | ||
GenBank mRNA | BC089802.1 | - | 5 | 351 | 7370033 | |
M55167.1 | - | 5 | 355 |