Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022507.1 | NP_071952.1 | 2, 3 | 348 | ||
XM_008764337.1 | XP_008762559.1 | 2, 3 | 249 | |||
GenBank mRNA | J04532.1 | - | 2, 3 | 348 |