Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004503.3 | NP_004494.1 | 1 | 294 | ||
NM_153693.4 | NP_710160.1 | 2 | 691 | |||
GenBank mRNA | AK314829.1 | - | 2 | 371 | ||
BC074844.2 | - | 1 | 208 | 30915460 | ||
BC074845.2 | - | 1 | 208 | 30915223 | ||
CR456954.1 | - | 1 | 182 | |||
M16938.1 | - | 2 | 418 | |||
S82986.1 | - | 1 | 314 |