Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022298.1 | NP_071634.1 | 4 | 760 | ||
GenBank mRNA | BC062238.1 | - | 4 | 794 | 6922126 | |
BC078830.1 | - | 4 | 773 | 7115494 | ||
FQ210110.1 | - | 4 | 792 | |||
FQ212756.1 | - | 4 | 726 | |||
FQ213186.1 | - | 4 | 792 | |||
FQ214446.1 | - | 2 | 349 | |||
FQ216135.1 | - | 1 | 207 | |||
FQ216180.1 | - | 1 | 207 | |||
V01227.1 | - | 4 | 760 |