Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022395.2 | NP_071790.1 | 6 | 696 | ||
GenBank mRNA | BC078826.1 | - | 6 | 691 | 7113892 | |
D13907.1 | - | 6 | 674 | |||
L12965.1 | - | 6 | 697 |