Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022505.2 | NP_071950.1 | 3, 4 | 529 | ||
XM_006239268.2 | XP_006239330.1 | 3, 4 | 563 | |||
GenBank mRNA | AB015191.1 | - | 3, 4 | 494 | ||
AF531096.1 | - | 3, 4 | 529 | |||
BC127471.1 | - | 3, 4 | 518 | 8374048 |