Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022515.1 | NP_071960.1 | 2, 3 | 96 | ||
GenBank mRNA | BC058473.1 | - | 2, 3 | 121 | 6918141 | |
FQ209984.1 | - | 2, 3 | 115 | |||
FQ220432.1 | - | 2, 3 | 125 | |||
FQ221142.1 | - | 2, 3 | 114 | |||
FQ221221.1 | - | 2, 3 | 121 | |||
FQ221486.1 | - | 2, 3 | 117 | |||
FQ221683.1 | - | 2, 3 | 108 | |||
FQ221688.1 | - | 2, 3 | 117 | |||
FQ221810.1 | - | 2, 3 | 117 | |||
FQ221946.1 | - | 2, 3 | 117 | |||
FQ221988.1 | - | 2, 3 | 121 | |||
FQ222823.1 | - | 2, 3 | 117 | |||
FQ223282.1 | - | 2, 3 | 117 | |||
FQ224575.1 | - | 2, 3 | 121 | |||
FQ224676.1 | - | 2, 3 | 117 | |||
X78443.1 | - | 2, 3 | 96 |