Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022522.2 | NP_071967.2 | 1, 2 | 82 | ||
GenBank mRNA | AF136231.1 | - | 1, 2 | 80 | ||
AF136232.1 | - | 1, 2 | 80 | |||
U77933.1 | - | 1, 2 | 82 |