Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022582.2 | NP_072104.2 | 1, 2 | 108 | ||
XM_006228668.2 | XP_006228730.1 | 4, 5 | 366 | |||
XM_006228670.2 | XP_006228732.1 | 3, 4 | 538 | |||
GenBank mRNA | AF036941.1 | - | 1, 2 | 117 | ||
U67883.1 | - | 2, 3 | 117 |