Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113740.1 | NP_001107212.1 | 3 | 275 | ||
NM_022616.2 | NP_072138.2 | 2 | 272 | |||
GenBank mRNA | AF142759.1 | - | 2 | 239 | ||
BC128738.1 | - | 3 | 173 | 7381331 | ||
CA512303.1 | - | 3 | 276 | |||
EV764168.1 | - | 3 | 247 |
Gene Symbol: LOC100910754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772553.1 | XP_008770775.1 | 1 | 264 |