Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022665.3 | NP_073156.3 | 3 | 271 | ||
XM_008767194.1 | XP_008765416.1 | 4 | 306 | |||
GenBank mRNA | X17611.1 | - | 3 | 270 |
Gene Symbol: Alppl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767355.1 | XP_008765577.1 | 4 | 2057 |