Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022668.2 | NP_073159.2 | 1, 2 | 178 | ||
XM_006255867.2 | XP_006255929.1 | 1, 2 | 651 | |||
XM_008772687.1 | XP_008770909.1 | 1, 2 | 655 | |||
GenBank mRNA | BC087717.1 | - | 1, 2 | 178 | 7317044 | |
FQ214199.1 | - | 1, 2 | 158 | |||
M99485.1 | - | 1, 2 | 178 |