Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022530.1 | NP_071975.1 | 2 | 198 | ||
GenBank mRNA | AF139808.1 | - | 2 | 198 | ||
FQ219962.1 | - | 2 | 220 | |||
FQ220048.1 | - | 2 | 219 | |||
FQ220059.1 | - | 2 | 221 | |||
FQ220802.1 | - | 2 | 221 | |||
FQ221033.1 | - | 2 | 220 | |||
FQ229694.1 | - | 2 | 219 | |||
FQ229792.1 | - | 2 | 220 | |||
FQ230178.1 | - | 2 | 221 |