Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022611.1 | NP_072133.1 | 4 | 536 | ||
XM_006246147.2 | XP_006246209.1 | 5 | 745 | |||
GenBank mRNA | AF133197.1 | - | 4 | 536 | ||
U16674.1 | - | 4 | 552 |