Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199849.1 | NP_001186778.1 | 5 | 411 | ||
NM_001199851.1 | NP_001186780.1 | 3 | 195 | |||
NM_004632.3 | NP_004623.1 | 4 | 315 | |||
NM_033657.2 | NP_387506.1 | 4 | 384 | |||
XM_005245481.2 | XP_005245538.1 | 3 | 260 | |||
XM_011509972.2 | XP_011508274.1 | 4 | 314 | |||
XM_017002289.1 | XP_016857778.1 | 4 | 383 | |||
XM_017002290.1 | XP_016857779.1 | 5 | 407 | |||
XM_017002292.1 | XP_016857781.1 | 4 | 280 | |||
XM_017002293.1 | XP_016857782.1 | 3 | 199 | |||
XM_017002295.1 | XP_016857784.1 | 4 | 280 | |||
XM_017002296.1 | XP_016857785.1 | 3 | 260 | |||
GenBank mRNA | AK225869.1 | - | 5 | 411 | ||
AK293117.1 | - | 4 | 316 | |||
AK298201.1 | - | 3 | 203 | |||
BC002358.1 | - | 3 | 343 | |||
BC107487.1 | - | 4 | 315 | 40032516 | ||
BC107488.1 | - | 4 | 384 | 40032512 | ||
BT019494.1 | - | 3 | 227 | |||
CR749790.1 | - | 4 | 395 | |||
U18321.1 | - | 4 | 396 | |||
X83544.1 | - | 4 | 300 |