Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022627.2 | NP_072149.1 | 2 | 204 | ||
XM_006233011.2 | XP_006233073.1 | 3 | 405 | |||
XM_006233012.2 | XP_006233074.1 | 3 | 305 | |||
GenBank mRNA | AF182717.1 | - | 2 | 206 | ||
BC078821.1 | - | 3 | 405 | 7135534 |