Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022633.1 | NP_072155.1 | 5 | 375 | ||
GenBank mRNA | AF188491.1 | - | 5 | 375 |