Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022521.3 | NP_071966.1 | 4 | 598 | ||
GenBank mRNA | BC061551.1 | - | 4 | 601 | 6890520 | |
M11842.1 | - | 3 | 541 |