Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270561.1 | NP_001257490.1 | 4 | 482 | ||
NM_001270562.1 | NP_001257491.1 | 4 | 380 | |||
NM_001270563.1 | NP_001257492.1 | 4 | 340 | |||
NM_022593.4 | NP_072115.1 | 4 | 376 | |||
XM_006237724.2 | XP_006237786.1 | 4 | 377 | |||
GenBank mRNA | BC060566.1 | - | 4 | 351 | 6918038 | |
CK355540.1 | - | 4 | 376 | |||
FM093546.1 | - | 4 | 342 | |||
FM101371.1 | - | 4 | 330 | |||
FM110992.1 | - | 4 | 346 | |||
FQ214805.1 | - | 4 | 482 | |||
L29259.1 | - | 3 | 293 |