Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mecp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022673.2 | NP_073164.2 | 3 | 315 | ||
XM_006229566.2 | XP_006229628.1 | 3 | 489 | |||
XM_006229567.2 | XP_006229629.1 | 2 | 599 | |||
GenBank mRNA | DQ897368.1 | - | 2 | 191 | ||
DQ897369.1 | - | 3 | 315 | |||
M94064.1 | - | 2 | 157 |