Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022686.2 | NP_073177.1 | 1 | 207 | ||
GenBank mRNA | AY936209.1 | - | 1 | 207 | ||
BC100619.1 | - | 1 | 211 | 7938313 |
Gene Symbol: Hist1h2ao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771770.1 | XP_008769992.1 | 1 | 216 |
Gene Symbol: Hist1h4m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271221.1 | NP_001258150.1 | 1, 2 | 188 |
Gene Symbol: Hist2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123469.1 | NP_001116941.1 | 1 | 217 |
Gene Symbol: LOC100910366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771686.1 | XP_008769908.1 | 4 | 1270 |
Gene Symbol: LOC102548682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771675.1 | XP_008769897.1 | 1 | 220 |
Gene Symbol: LOC502940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY936210.1 | - | 1 | 193 |
Gene Symbol: LOC680097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001055696.4 | XP_001055696.1 | 1 | 239 |