Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022698.1 | NP_073189.1 | 2 | 284 | ||
XM_006230896.2 | XP_006230958.1 | 2 | 989 | |||
XM_006230897.2 | XP_006230959.1 | 1 | 167 | |||
XM_006230898.2 | XP_006230960.1 | 2 | 182 | |||
GenBank mRNA | AF003523.1 | - | 2 | 284 | ||
AF279910.1 | - | 2 | 284 | |||
AF279911.1 | - | 2 | 284 | |||
BC061728.1 | - | 2 | 529 | 5598682 |